• USA Home
  • Search Results

  • bis2ethylhexylphthalate3905611781711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(2-ethylhexyl) phthalate

3 Products
Synonym:   ‘Dioctyl’ phthalate, DEHP, Phthalic acid bis(2-ethylhexyl ester)
Image of Bis(2-ethylhexyl) phthalate
CAS Number:
Empirical Formula (Hill Notation):
 C24H38O4
Molecular Weight:
 390.56
 Beilstein/REAXYS Number:
 1890696
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Shelf Life
  Product Line
  Physical Form
  Special Grade
Show More