• USA Home
  • Search Results

  • bis2ethylhexylsebacate4266712262311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(2-ethylhexyl) sebacate

4 Products
Synonym:   ‘Dioctyl’ sebacate, Di(2-ethylhexyl) sebacate, Sebacic acid di(2-ethylhexyl) ester
Image of Bis(2-ethylhexyl) sebacate
CAS Number:
Linear Formula:
 [-(CH2)4CO2CH2CH(C2H5)(CH2)3CH3]2
Molecular Weight:
 426.67
 Beilstein/REAXYS Number:
 1806504
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More