• USA Home
  • Search Results

  • bis2hydroxyphenylmethane20023246702911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(2-hydroxyphenyl)methane

1 Products
Synonym:   2,2′-Methylenediphenol
Image of Bis(2-<WBR>hydroxyphenyl)<WBR>methane
CAS Number:
Linear Formula:
 CH2(C6H4OH)2
Molecular Weight:
 200.23
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More