• USA Home
  • Search Results

  • bisbenzimideh33258533882349145411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

bisBenzimide H 33258

3 Products
Synonym:   2′-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi(1H-benzimidazole) trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazoyl]-6-(1-methyl-4-piperazyl)benzimidazole trihydrochloride, BBIH, BXI-72, HOE 33258, Hoechst 33258, NSC334072
Image of bisBenzimide H 33258
CAS Number:
Empirical Formula (Hill Notation):
 C25H24N6O · 3HCl
Molecular Weight:
 533.88
 Beilstein Registry Number:
 4088183
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Suitability
  Fluorescence Test
  Physical Form
  Assay
Show More