• USA Home
  • Search Results

  • bisbenzonitrilepalladiumiichloride383571422064511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(benzonitrile)palladium(II) chloride

1 Products
Synonym:   Benzonitrile, palladium complex, Bis(benzonitrile)dichloropalladium(II), Bis(benzonitrile)palladium(II) chloride, Bis(phenylnitrile)dichloropalladium, Dibenzonitrilepalladium dichloride, Dichlorobis(benzonitrile)palladium(II), Dichlorobis(benzonitrile)palladium, Dichlorobis(phenyl cyanide)palladium, Palladium dichloride bis(benzonitrile), Palladium(II) chloride bis(benzonitrile) complex
Image of Bis(benzonitrile)<WBR>palladium(II) chloride
CAS Number:
Linear Formula:
 (C6H5CN)2PdCl2
Molecular Weight:
 383.57
 Beilstein/REAXYS Number:
 3981730
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More