• USA Home
  • Search Results

  • biscyclopentadienylmagnesiumii15449128472611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(cyclopentadienyl)magnesium(II)

1 Products
Synonym:   Bis(cyclopentadienyl)magnesium
Image of Bis(cyclopentadienyl)<WBR>magnesium(II)
CAS Number:
Linear Formula:
 Mg(C5H5)2
Molecular Weight:
 154.49
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More