• USA Home
  • Search Results

  • bisethylcyclopentadienylnickelii244993188651811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(ethylcyclopentadienyl)nickel(II)

1 Products
Image of Bis(ethylcyclopentadienyl)<WBR>nickel(II)
CAS Number:
Linear Formula:
 Ni(C5H4C2H5)2
Molecular Weight:
 244.99
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More