• USA Home
  • Search Results

  • bismethylthiomethane10823161826411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(methylthio)methane

2 Products
Synonym:   Dimethylthiomethane, Formaldehyde dimethyl mercaptal, Truffle sulfide
Image of Bis(methylthio)<WBR>methane
CAS Number:
Linear Formula:
 CH3SCH2SCH3
Molecular Weight:
 108.23
 Beilstein/REAXYS Number:
 1731143
 EC Number:
 FEMA Number:
 3878
 Flavis number:
 12.118
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Assay
Show More