• USA Home
  • Search Results

  • bismuthiiicitrate3980881393411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bismuth(III) citrate

1 Products
Synonym:   Bismuth citrate
Image of Bismuth(III) citrate
CAS Number:
Linear Formula:
 [O2CCH2C(OH)(CO2)CH2CO2]Bi
Molecular Weight:
 398.08
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More