• USA Home
  • Search Results

  • bismuthiiifluoride26598778761311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bismuth(III) fluoride

1 Products
Synonym:   Trifluorobismuthine
Image of Bismuth(III) fluoride
CAS Number:
Empirical Formula (Hill Notation):
 BiF3
Molecular Weight:
 265.98
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More