• USA Home
  • Search Results

  • bisoctrizole6588710359745111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bisoctrizole

3 Products
Synonym:   2,2′-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol]
Image of Bisoctrizole
CAS Number:
Empirical Formula (Hill Notation):
 C41H50N6O2
Molecular Weight:
 658.87
 Beilstein/REAXYS Number:
 11326835
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More