• USA Home
  • Search Results

  • bisphenola228298005711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bisphenol A

4 Products
Synonym:   2,2-Bis(4-hydroxyphenyl)propane, 4,4′-Isopropylidenediphenol
Image of Bisphenol A
CAS Number:
Linear Formula:
 (CH3)2C(C6H4OH)2
Molecular Weight:
 228.29
 Beilstein/REAXYS Number:
 1107700
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More