• USA Home
  • Search Results

  • bisphenols250278009111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bisphenol S

1 Products
Synonym:   4,4′-Sulfonyldiphenol, 4-Hydroxyphenyl sulfone, Bis(4-hydroxyphenyl) sulfone, Bisphenol S
Image of Bisphenol S
CAS Number:
Linear Formula:
 O2S(C6H4OH)2
Molecular Weight:
 250.27
 Beilstein/REAXYS Number:
 2052954
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More