• USA Home
  • Search Results

  • bistrifluoroacetoxyiodobenzene43004271278911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

[Bis(trifluoroacetoxy)iodo]benzene

2 Products
Synonym:   BTI, Iodobenzene I,I-bis(trifluoroacetate), PIFA
Image of [Bis(trifluoroacetoxy)<WBR>iodo]<WBR>benzene
CAS Number:
Linear Formula:
 (CF3CO2)2IC6H5
Molecular Weight:
 430.04
 Beilstein/REAXYS Number:
 764767
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More