• USA Home
  • Search Results

  • bistrimethylsilylacetylene170401463040111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(trimethylsilyl)acetylene

1 Products
Synonym:   BTMSA
Image of Bis(trimethylsilyl)<WBR>acetylene
CAS Number:
Linear Formula:
 (CH3)3SiC≡CSi(CH3)3
Molecular Weight:
 170.40
 Beilstein/REAXYS Number:
 906870
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More