• USA Home
  • Search Results

  • bistriphenylphosphinerhodiumicarbonylchloride690941393894811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(triphenylphosphine)rhodium(I) carbonyl chloride

1 Products
Synonym:   Chlorocarbonylbis(triphenylphosphine)rhodium(I), RhCl(CO)(PPh3)2, Rhodiumbis(triphenylphosphine) carbonyl chloride
Image of Bis(triphenylphosphine)<WBR>rhodium(I) carbonyl chloride
CAS Number:
Linear Formula:
 [(C6H5)3P]2RhCl(CO)
Molecular Weight:
 690.94
 Beilstein/REAXYS Number:
 4618896
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More