• USA Home
  • Search Results

  • bistrispropane282336443196511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

BIS-TRIS propane

5 Products
Synonym:   1,3-Bis[tris(hydroxymethyl)methylamino]propane
Image of BIS-TRIS propane
CAS Number:
Linear Formula:
 CH2[CH2NHC(CH2OH)3]2
Molecular Weight:
 282.33
 Beilstein/REAXYS Number:
 1786109
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  pH Value
  Application(s)
  Absorption
  Solubility
  Product Line
  Physical Form
Show More