• USA Home
  • Search Results

  • biuret1030810819011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Biuret

1 Products
Synonym:   Allophanic acid amide, Carbamoylurea
Image of Biuret
CAS Number:
Linear Formula:
 NH2CONHCONH2
Molecular Weight:
 103.08
 Beilstein/REAXYS Number:
 1703510
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More