• USA Home
  • Search Results

  • blactamasefromenterobactercloacae12345907360311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Lactamase from Enterobacter cloacae

2 Products
Synonym:   β-Lactamase I, β-Lactamase II, Cephalosporinase, Penicillin amido-β-lactam hydrolase, Penicillinase from Enterobacter cloacae
CAS Number:
 Enzyme Commission (EC) Number:
 3.5.2.6  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More