• USA Home
  • Search Results

  • bleomycinsulfatefromstreptomycesverticillus12345904193411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bleomycin sulfate from Streptomyces verticillus

4 Products
Synonym:   Blenoxane, Bleo, Blexane
Image of Bleomycin sulfate from <I>Streptomyces verticillus</I>
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.76
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Special Grade
  Application(s)
  Product Line
  Color
  Solubility
  Mode of Action
  Physical Form
Show More