• USA Home
  • Search Results

  • bnacetylglucosaminidasefromcanavaliaensiformisjackbean12345901233311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-N-Acetylglucosaminidase from Canavalia ensiformis (Jack bean)

1 Products
Synonym:   β-N-Acetyl-D-hexosaminide N-acetylhexosaminohydrolase, β-N-Acetylhexosaminidase
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.2.1.52  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More