• USA Home
  • Search Results

  • bnicotinamideadeninedinucleotide2phosphatereducedtetrasodiumsalthydrate83335anhydrousbasis2646711anhydrous11

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate

4 Products
Synonym:   β-NADPH, 2′-NADPH hydrate, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt
Image of β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C21H26N7Na4O17P3 · xH2O
Molecular Weight:
 833.35 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More