• USA Home
  • Search Results

  • bnicotinamideadeninedinucleotidephosphatesodiumsalt76539118416311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Nicotinamide adenine dinucleotide phosphate sodium salt

2 Products
Synonym:   β-NADP-Na, β-NADP-sodium salt, Coenzyme II sodium salt, NADP, TPN-Na, TPN, Triphosphopyridine nucleotide sodium salt
Image of β-Nicotinamide adenine dinucleotide phosphate sodium salt
CAS Number:
Linear Formula:
 C21H27N7O17P3 · Na
Molecular Weight:
 765.39
 Beilstein/REAXYS Number:
 4779954
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More