• USA Home
  • Search Results

  • bocaiboh203243099229111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-Aib-OH

2 Products
Synonym:   α-(Boc-amino)isobutyric acid, Boc-α-methylalanine
Image of Boc-Aib-OH
CAS Number:
Linear Formula:
 (CH3)3CO2CNHC(CH3)2CO2H
Molecular Weight:
 203.24
 Beilstein/REAXYS Number:
 1953772
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More