• USA Home
  • Search Results

  • bocargoh274321372676611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-Arg-OH

1 Products
Synonym:   Nα-(tert-Butoxycarbonyl)-L-arginine, Nα-Boc-L-arginine
Image of Boc-Arg-OH
CAS Number:
Linear Formula:
 HN=C(NH2)NH(CH2)3CH(COOH)NHC(O)OC(CH3)3
Molecular Weight:
 274.32
 Beilstein/REAXYS Number:
 2419628
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
408484 Boc-Arg-OH,