• USA Home
  • Search Results

  • bocbiodoalaobzl4052310895720611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-β-iodo-Ala-OBzl

1 Products
Synonym:   N-(tert-Butoxycarbonyl)-3-iodo-L-alanine benzyl ester, Boc-3-iodo-L-alanine benzyl ester
Image of Boc-β-iodo-Ala-OBzl
CAS Number:
Linear Formula:
 ICH2CH[NHCO2C(CH3)3]CO2CH2C6H5
Molecular Weight:
 405.23
 Beilstein/REAXYS Number:
 4506658
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
406252 Boc-β-iodo-Ala-OBzl, 97%,