• USA Home
  • Search Results

  • bocbphenylpheoh3414013866263211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-β-phenyl-Phe-OH

1 Products
Synonym:   N-(tert-Butoxycarbonyl)-β-phenyl-L-phenylalanine, Boc-β,β-diphenyl-Ala-OH, Boc-3,3-diphenyl-L-alanine
Image of Boc-β-phenyl-Phe-OH
CAS Number:
Linear Formula:
 (C6H5)2CHCH[NHCO2C(CH3)3]CO2H
Molecular Weight:
 341.40
 Beilstein/REAXYS Number:
 4880641
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More