• USA Home
  • Search Results

  • bocchaome285389810541011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-Cha-OMe

1 Products
Synonym:   N-(tert-Butoxycarbonyl)-3-cyclohexyl-L-alanine methyl ester, Boc-3-cyclohexyl-L-alanine methyl ester
Image of Boc-Cha-OMe
CAS Number:
Linear Formula:
 C6H11CH2CH[NHCO2C(CH3)3]CO2CH3
Molecular Weight:
 285.38
 Beilstein/REAXYS Number:
 5057730
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
421715 Boc-Cha-OMe, 98%,