• USA Home
  • Search Results

  • bocd1naloh315367693248411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-D-1-Nal-OH

1 Products
Synonym:   Boc-3-(1-naphthyl)-D-alanine
Image of Boc-<SC>D</SC>-1-Nal-OH
CAS Number:
Linear Formula:
 C10H7CH2CH[NHCO2C(CH3)3]CO2H
Molecular Weight:
 315.36
 Beilstein/REAXYS Number:
 6180114
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More