• USA Home
  • Search Results

  • bocdgluobzl337373440430311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-D-Glu-OBzl

2 Products
Synonym:   Boc-D-glutamic acid 1-benzyl ester
Image of Boc-<SC>D</SC>-Glu-OBzl
CAS Number:
Empirical Formula (Hill Notation):
 C17H23NO6
Molecular Weight:
 337.37
 Beilstein/REAXYS Number:
 4154458
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More