• USA Home
  • Search Results

  • bocglnoh246261372685711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-Gln-OH

2 Products
Synonym:   Nα-(tert-Butoxycarbonyl)-L-glutamine, Boc-L-glutamine
Image of Boc-Gln-OH
CAS Number:
Linear Formula:
 NH2COCH2CH2CH(COOH)NHCOOC(CH3)3
Molecular Weight:
 246.26
 Beilstein/REAXYS Number:
 2127805
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More