• USA Home
  • Search Results

  • bochistosoh409463589943511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-His(Tos)-OH

2 Products
Synonym:   N-α-t.-Boc-N-im-tosyl-L-histidine
Image of Boc-His(Tos)-OH
CAS Number:
Linear Formula:
 C₁₈H₂₃N₃O₆S
Molecular Weight:
 409.46 g/mol
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More