• USA Home
  • Search Results

  • bochypoh231251372669711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-Hyp-OH

1 Products
Synonym:   trans-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline, Boc-L-hydroxyproline
Image of Boc-Hyp-OH
CAS Number:
Empirical Formula (Hill Notation):
 C10H17NO5
Molecular Weight:
 231.25
 Beilstein/REAXYS Number:
 4295484
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
15544 Boc-Hyp-OH, ≥98.0% (TLC),