• USA Home
  • Search Results

  • boclyszosu477513440436911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-Lys(Z)-OSu

1 Products
Synonym:   Nα-Boc-Nε-Cbz-L-lysine N-hydroxysuccinimide ester, Nα-Boc-Nε-Z-L-lysine hydroxysuccinimide ester, Nε-Z-Nα-Boc-L-lysine hydroxysuccinimide ester
Image of Boc-Lys(Z)-OSu
CAS Number:
Empirical Formula (Hill Notation):
 C23H31N3O8
Molecular Weight:
 477.51
 Beilstein/REAXYS Number:
 1559393
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
15541 Boc-Lys(Z)-OSu,