• USA Home
  • Search Results

  • bocosu215201313912311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-OSu

1 Products
Synonym:   N-(tert-Butoxycarbonyloxy)succinimide, tert-Butyl N-succinimidyl carbonate
Image of Boc-OSu
CAS Number:
Empirical Formula (Hill Notation):
 C9H13NO5
Molecular Weight:
 215.20
 Beilstein/REAXYS Number:
 1536756
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
20430 Boc-OSu, ≥98.0% (CHN),