• USA Home
  • Search Results

  • bocpropargylglyoh213236303948511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boc-propargyl-Gly-OH

1 Products
Synonym:   (S)-2-(Boc-amino)-4-pentynoic acid, Boc-L-2-propargylglycine, Boc-Pra-OH
Image of Boc-propargyl-Gly-OH
CAS Number:
Empirical Formula (Hill Notation):
 C10H15NO4
Molecular Weight:
 213.23
 Beilstein/REAXYS Number:
 5739897
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More