• USA Home
  • Search Results

  • boricacid61831004335311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boric acid

22 Products
Image of Boric acid
CAS Number:
Linear Formula:
 H3BO3
Molecular Weight:
 61.83
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss:
 38120104
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  Anal. Standard Form
  Suitability
  Loss
  Restriction
  Purity Grade
  Package quantity
  Manufacturer designation
  Application(s)
  Density
  Bulk Density
  Product Line
  Physical Form
  UV-Absorption Values
  Potency
  Agency/Method
  Solubility
  pH Value
  Special Grade
Show More

Product #
Description
B6768 Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%,
B0394 Boric acid, ACS reagent, ≥99.5%,
PHR2664 Boric acid, Pharmaceutical Secondary Standard; Certified Reference Material, New
1.00165 Boric acid, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
B7901 Boric acid, for electrophoresis, ≥99.5%,
B0252 Boric acid, ReagentPlus®, ≥99.5%,
31146 Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%,
202878 Boric acid, 99.999% trace metals basis,
B7660 Boric acid, BioXtra, ≥99.5%,
15663 Boric acid, BioUltra, for molecular biology, ≥99.5% (T),
339067 Boric acid, 99.97% trace metals basis,
B2645 Boric acid, tablet, 1 g boric acid per tablet,
1.00765 Boric acid, 99.9999 Suprapur®,
11607 Boric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder,
1.00160 Boric acid, cryst., EMPROVE® ESSENTIAL Ph Eur,BP,NF,
1.00162 Boric acid, powder, EMPROVE® ESSENTIAL Ph Eur,BP,NF,
BX0865 Boric acid, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
1.00156 Boric acid, cryst., EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF,
1.00164 Boric acid, cryst., EMPROVE® API Ph Eur,BP,NF,
NIST973 Boric acid, NIST® SRM® 973, acidimetric standard,
1076341 Boric acid, United States Pharmacopeia (USP) Reference Standard,
NIST951A Boric acid, NIST® SRM® 951a, isotopic standard,