• USA Home
  • Search Results

  • borontrichloridesolution117171029434511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boron trichloride solution

6 Products
Image of Boron trichloride solution
CAS Number:
Empirical Formula (Hill Notation):
 BCl3
Molecular Weight:
 117.17
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Vapor pressure
  Density
  Reaction Suitability
Show More