• USA Home
  • Search Results

  • borontrifluoridediethyletherate1419310963711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boron trifluoride diethyl etherate

3 Products
Synonym:   Boron trifluoride ethyl etherate
Image of Boron trifluoride diethyl etherate
CAS Number:
Linear Formula:
 BF3 · O(C2H5)2
Molecular Weight:
 141.93
 Beilstein/REAXYS Number:
 3909607
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
  Special Grade
  Assay
Show More