• USA Home
  • Search Results

  • borontrifluorideethylaminecomplex112897523011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boron trifluoride ethylamine complex

1 Products
Image of Boron trifluoride ethylamine complex
CAS Number:
Linear Formula:
 BF3 · C2H5NH2
Molecular Weight:
 112.89
 Beilstein/REAXYS Number:
 3672802
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More