• USA Home
  • Search Results

  • borontrifluoridemethanolsolution998537357911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Boron trifluoride-methanol solution

3 Products
Synonym:   Trifluoroborane
Image of Boron trifluoride-methanol solution
CAS Number:
Linear Formula:
 BF3 · MeOH
Molecular Weight:
 99.85
 Beilstein/REAXYS Number:
 3611499
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
  Application(s)
  Assay
  Physical Form
  Special Grade
  Density
  Concentration
Show More