• USA Home
  • Search Results

  • bovineserumalbumin12345904846811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bovine serum albumin

43 Products
Synonym:   Albumin bovine serum, BSA, Bovine Serum Albumin, Bovine albumin
Image of Bovine serum albumin
CAS Number:
 EC Number:
 MDL number:
 NACRES
 NA.27
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  pH Value
  Restriction
  Package quantity
  Special Grade
  Quality clarifier
  Molecular weight
  Product Line
  Suitability
  Loss
  Geographic origin
  Purification method
  Assay
  Solubility
  Biological Source
  Physical Form
Show More

Product #
Description
A8531 Bovine Serum Albumin, pH <5.0, powder,
A0281 Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis),
A3912 Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%,
A3311 Bovine Serum Albumin, powder, BioXtra,
A2058 Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture,
A4919 Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%,
A5611 Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis),
A7511 Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized,
B6917 Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated,
A4161 Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture,
A4378 Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis),
A1933 Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%,
A7517 Bovine Serum Albumin, For use as a marker in SDS-PAGE,
A1470 Bovine Serum Albumin, low endotoxin, lyophilized powder, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis),
05470 Bovine serum albumin, lyophilized powder, crystallized, ≥98.0% (GE),
A9430 Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%,
A3156 Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture,
A8654 Bovine Serum Albumin, dimer mol wt ~132 kDa,
A3858 Bovine Serum Albumin, lyophilized powder, Protease, essentially free, ≥98% (agarose gel electrophoresis),
A2934 Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis),
A9085 Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis),
A3983 Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis),
A3733 Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis),
A9543 Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%,
A9056 Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis),
A3608 Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis),
A4737 Bovine Serum Albumin, lyophilized powder, protease, essentially free, ≥98% (agarose gel electrophoresis),
A4612 Bovine Serum Albumin, lyophilized powder, Essentially fatty acid free,
A8022 Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%,
A7638 Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis),
A3803 Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis),
A7888 Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%,
A8806 Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis),
B4287 Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%,
A3294 Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%,
A6003 Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis),
A9418 Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture,
A9647 Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%,
A3059 Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%,
A4503 Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%,
A7030 Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%,
A2153 Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis),
A7906 Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%,