• USA Home
  • Search Results

  • bovineserumalbuminsolution12345904846811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bovine Serum Albumin solution

18 Products
Synonym:   Albumin solution from bovine serum, BSA solution
Image of Bovine Serum Albumin solution
CAS Number:
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Additive
  Quality clarifier
  ID clarifier
  Suitability
  Product Line
  Purification method
  Geographic origin
  Sterilization
  Physical Form
  Concentration
  Biological Source
Show More

Product #
Description
A9576 Bovine Serum Albumin solution, 30% in DPBS, sterile-filtered, BioXtra, suitable for cell culture,
A8412 Bovine Serum Albumin solution, 7.5% in DPBS, sterile-filtered, BioXtra, suitable for cell culture,
A7979 Bovine Serum Albumin solution, 35% in DPBS, sterile-filtered, BioXtra, suitable for cell culture,
A7284 Bovine Serum Albumin solution, 30%±2% in 0.85% sodium chloride, aseptically filled,
B8667 Bovine Serum Albumin solution, 20 mg/mL in H2O, low bioburden, protease free, for molecular biology,
A1595 Bovine Serum Albumin solution, 10% in DPBS, low endotoxin, fatty acid free, suitable for cell culture, sterile-filtered,
A7409 Bovine Serum Albumin solution, 35% in 0.85% sodium chloride, aseptically filled,
A9205 Bovine Serum Albumin solution, 30% in saline, fatty acid free, aseptically filled,
A4628 Bovine Serum Albumin solution, 5% in 0.85% sodium chloride, aseptically filled,
A8577 Bovine Serum Albumin solution, 30% in saline, protease free, aseptically filled,
A0336 Bovine Serum Albumin solution, 30% in saline, IgG free, aseptically filled,
A7159 Bovine Serum Albumin solution, 25% in Tyrode's buffer, aseptically filled,
A7034 Bovine Serum Albumin solution, 22% in saline, contains azide, aseptically filled,
A8327 Bovine Serum Albumin solution, 30%±2% in 0.85% sodium chloride, aseptically filled,
A7534 Bovine Serum Albumin solution, 35% in saline, contains azide, aseptically filled,
A1662 Bovine Serum Albumin solution, 30%±2% in 0.85% sodium chloride, aseptically filled,
A3299 Bovine Serum Albumin solution, 30% in 0.85% sodium chloride, aseptically filled,
A3174 Bovine Serum Albumin solution, 30% in saline, high avidity, contains azide and caprylate, aseptically filled,