• USA Home
  • Search Results

  • bpropiolactone72065757811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Propiolactone

1 Products
Synonym:   3-Hydroxypropionic acid lactone, Hydracrylic acid β-lactone, Oxetan-2-one
Image of β-Propiolactone
CAS Number:
Empirical Formula (Hill Notation):
 C3H4O2
Molecular Weight:
 72.06
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More