• USA Home
  • Search Results

  • bq1236326813655381611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

BQ-123

1 Products
Image of BQ-123
CAS Number:
Empirical Formula (Hill Notation):
 C31H41N6O7 Na
Molecular Weight:
 632.68
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More