• USA Home
  • Search Results

  • brefeldina280362035015611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Brefeldin A

3 Products
Synonym:   γ,4-Dihydroxy-2-(6-hydroxy-1-heptenyl)-4-cyclopentanecrotonic acid λ-lactone, Ascotoxin, BFA, Cyanein, Decumbin
Image of Brefeldin A
CAS Number:
Empirical Formula (Hill Notation):
 C16H24O4
Molecular Weight:
 280.36
 Beilstein Registry Number:
 25191
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Physical Form
  Assay
  Mode of Action
  Solubility
Show More