• USA Home
  • Search Results

  • brij5812345900495911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Brij® 58

2 Products
Synonym:   Brij® 58, Polyoxyethylene (20) cetylether, Polyethyleneglycol (20) cetylether
Image of Brij<SUP>®</SUP> 58
CAS Number:
Linear Formula:
 HO-(CH2CH2O)n-(CH2)15-CH3
 EC Index Number
 500-014-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Melting Point
  Transition temperature
  Molecular weight
Show More