• USA Home
  • Search Results

  • brijl2312345900292011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Brij® L23

2 Products
Synonym:   Brij 35, C12E23, Polyoxyethylene (23) lauryl ether
Image of Brij<SUP>®</SUP> L23
CAS Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Critical micelle concentration
  Titertest
  Molecular weight
  ID clarifier
Show More