• USA Home
  • Search Results

  • brijs2012345900500911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Brij® S20

1 Products
Synonym:   Polyethylene glycol octadecyl ether, Polyoxyethylene (20) stearyl ether
Image of Brij<SUP>®</SUP> S20
CAS Number:
Linear Formula:
 C18H37(OCH2CH2)nOH, n~20
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
P4019 Brij® S20,