• USA Home
  • Search Results

  • brilliantblueg85402610458111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Brilliant Blue G

2 Products
Synonym:   Acid blue 90, Coomassie Brilliant Blue G
Image of Brilliant Blue G
CAS Number:
Empirical Formula (Hill Notation):
 C47H48N3NaO7S2
Molecular Weight:
 854.02
 Beilstein/REAXYS Number:
 5230822
 Colour Index Number:
 42655
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More